Learn More
Bochem™ Thermometer Clamp
Product Code. 10478432
Change view
Click to view available options
Product Type:
Thermometer Clamp
Unit Size:
Each
Product Code. Product Type Unit Size
10478432 Thermometer Clamp Each
1 options
Product Code. 10478432 Supplier Bochem™ Supplier No. 5621
Beräknad leverans: 17-12-2026
Designed with a swiveling bosshead for versatile, 360° positioning. Bochem™ Thermometer Clamp has a screw clamp and v-notch rod-holding block and independently adjustable jaws.
  • Swiveling bosshead.
  • V-notch and screw clamp for support rod positioning.
  • Independently adjustable jaws.
Specifications
Weight (Metric) 119 g
Product Type Thermometer Clamp