Läs mer
Chromium dioxide, TRC
Produktkod. 30362188
Change view
Kvantitet:
10 g
Produktkod. Kvantitet
30362188 10 g
1 options
Produktkod. 30362188 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC27411010G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Chromium dioxide
CAS 12018-01-8
Molekylformel CrO2
Kvantitet 10 g
Synonym Chromium oxide (CrO2), Amdry 6415, Chromium dioxide, Chromium dioxide (CrO2), Chromium oxide, Chromium(IV) oxide, Magtrieve
InChI formel InChI=1S/Cr.2O
LEDER O=[Cr]=O
IUPAC-namn dioxochromium
Molekylvikt (g/mol) 83.99
Formel vikt 83.9303