missing translation for 'onlineSavingsMsg'
Learn More

Chromium(III) Oxide, ≥98%, Honeywell Fluka™

Catalog Number p-7120325
Click to view available options
Quantity:
100 g
500 g
2 kg
Packaging:
Poly Bottle
This item is not returnable. View return policy
1308-38-9
Cr2O3
151.99
MFCD00010949
UOUJSJZBMCDAEU-UHFFFAOYSA-N
chromium iii oxide, chromia, dichromium trioxide, chromium sesquioxide, chrome green, green chromium oxide, chromium 3+ oxide, green cinnabar, chrome ochre, chrome oxide
517277
CHEBI:48242
[O--].[O--].[O--].[Cr+3].[Cr+3]
This item is not returnable. View return policy
1308-38-9
Cr2O3
151.99
MFCD00010949
UOUJSJZBMCDAEU-UHFFFAOYSA-N
chromium iii oxide, chromia, dichromium trioxide, chromium sesquioxide, chrome green, green chromium oxide, chromium 3+ oxide, green cinnabar, chrome ochre, chrome oxide
517277
CHEBI:48242
[O--].[O--].[O--].[Cr+3].[Cr+3]

Powder,≥98%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY
Boiling Point 4000°C
Melting Point 2435°C
Color Green
Quantity 100 g
UN Number NONH for all modes of transport
IUPAC Name oxo(oxochromiooxy)chromium
Formula Weight 151.99g/mol
Percent Purity ≥98%
Packaging Poly Bottle
Appearance Powder
Density 5.2g/cm3
Chemical Name or Material Chromium(III) oxide
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'