missing translation for 'onlineSavingsMsg'
Learn More
Chromium, plasma standard solution, Specpure™ Cr 1000μg/mL Product Code.: 11341439

Chromium, plasma standard solution, Specpure™ Cr 1000μg/mL

Product Code. p-4408540
50 mL, 50mL
Click to view available options
Quantity:
50 mL
100 mL
500 mL
Unit Size:
100mL
500mL
50mL
This item is not returnable. View return policy

Product Code. 11341439

Brand: Thermo Scientific Alfa Aesar 013864.AD

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7440-47-3 | Cr | 52.00 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7440-47-3, 7647-01-0, 7732-18-5
Molecular Formula Cr
Molecular Weight (g/mol) 52.00
MDL Number MFCD00010944
InChI Key VYZAMTAEIAYCRO-UHFFFAOYSA-N
PubChem CID 23976
ChEBI CHEBI:28073
IUPAC Name chromium
SMILES [Cr]

Specifications

Vapor Pressure 23 hPa (17mm Hg) at 20°C
Physical Form Liquid
Color Colorless
Quantity 50 mL
Concentration or Composition (by Analyte or Components) Hydrochloric acid: 5.0%; Chromium: 0.1%; Water: 94.9%
Chemical Name or Material Chromium

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Product Code
Lot Number1550390 Certificate TypeCertificate of Analysis Date10/04/2025 Product Code013864.AD,013864.AE,013864.AP
Lot Number1521865 Certificate TypeCertificate of Analysis Date13/03/2025 Product Code013864.AD,013864.AE,013864.AP
Lot Number1521866 Certificate TypeCertificate of Analysis Date23/01/2025 Product Code013864.AD,013864.AE,013864.AP
Lot Number1517325 Certificate TypeCertificate of Analysis Date15/01/2025 Product Code013864.AD,013864.AE,013864.AP
Lot Number1517324 Certificate TypeCertificate of Analysis Date15/01/2025 Product Code013864.AD,013864.AE,013864.AP

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Chromium, plasma standard solution, Specpure™ Cr 1000μg/mL >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.