Läs mer
Chromium (VI) Standard: Cr+6 @ 1000 μg/mL in H2O, VHG™
Produktkod. 18032750
Change view
Kvantitet:
50 ml
100 ml
250 ml
500 ml
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
18032750 500 ml
18060070 250 ml
18012450 50 ml
18021450 100 ml
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Produktkod. 18032750 Leverantör VHG Labs Leverantörsnummer VHGPCR6W500
Vänligen för att beställa denna produkten. Behöver du ett Webbkonto? Registrera ditt konto idag!
VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications
Chromium (VI) Standard: Cr+6 @ 1000 μg/mL in H2O
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Chromium+6
Metoder Speciation
Analyt- eller komponentnamn Chromium+6
Frakt skick Room Temperature
Molekylformel Cr6+
Synonym Cr6+
LEDER [Cr6+]
IUPAC-namn Chromium+6
Lösningsmedel eller matris Water
Molekylvikt (g/mol) 52
Kvantitet 500 ml
Formel vikt 52 g/mol