Läs mer
Chromonar Hydrochloride, TRC
Produktkod. 30356799
Change view
Kvantitet:
10 mg
25 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30356799 10 mg
30353765 100 mg
30350976 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30356799 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC43268510MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 655-35-6
Molekylformel C20 H27 N O5 . Cl H
Molekylvikt (g/mol) 397.89
Synonym Acetic acid, 2-[[3-[2-(diethylamino)ethyl]-4-methyl-2-oxo-2H-1-benzopyran-7-yl]oxy]-, ethyl ester, hydrochloride (1:1), Acetic acid, [[3-[2-(diethylamino)ethyl]-4-methyl-2-oxo-2H-1-benzopyran-7-yl]oxy]-, ethyl ester, hydrochloride (7CI,8CI,9CI), 3-(2-Diethylaminoethyl)-4-methyl-7-ethoxycarbonylmethoxycoumarin hydrochloride, 3-(β-Diethylaminoethyl)-4-methyl-7-carbethoxymethoxy-2-oxo-(1,2-chromene) hydrochloride, 3-(β-Diethylaminoethyl)-4-methyl-7-carbethoxymethoxy-2-oxo-2H-1-benzopyran hydrochloride, 3-[2-(Diethylamino)ethyl]-4-methyl-7-(carbethoxymethoxy)coumarin hydrochloride, 3-[2-(Diethylamino)ethyl]-4-methyl-7-carbethoxymethoxy-2-oxo-2H-chromen hydrochloride, A 27053, AG 3, Antiangor, Carbochromen hydrochloride, Carbochromene hydrochloride, Carbocromen hydrochloride, Carbocromene hydrochloride, Casella 4489, Cassella 4489, Chromonar hydrochloride, Ethyl 3-(β-diethylaminoethyl)-4-methylcoumarin-7-oxyacetate hydrochloride, Ethyl [[3-[2-(diethylamino)ethyl]-4-methyl-2-oxo-2H-1-benzopyran-7-yl]oxy]acetate hydrochloride, Intenkordin, Intenkordin hydrochloride, Intensain, Intensain hydrochloride, Intercordin, Interkordin, NSC 110430
IUPAC-namn ethyl 2-[3-[2-(diethylamino)ethyl]-4-methyl-2-oxochromen-7-yl]oxyacetate;hydrochloride
LEDER Cl.CCOC(=O)COc1ccc2C(=C(CCN(CC)CC)C(=O)Oc2c1)C
Specifikationer
Kemiskt namn eller material Chromonar hydrochloride
Kvantitet 10 mg
InChI formel InChI=1S/C20H27NO5.ClH/c1-5-21(6-2)11-10-17-14(4)16-9-8-15(12-18(16)26-20(17)23)25-13-19(22)24-7-3;/h8-9,12H,5-7,10-11,13H2,1-4H3;1H
Formel vikt 397.1656