Learn More
Proteintech ChromoTek RFP-Trap™ Magnetic Particles M-270
RFP-Trap™ Magnetic Particles M-270 for immunoprecipitation (IP) of RFP-tagged proteins.
Brand: Proteintech RTD-20
Additional Details : Weight : 0.00240kg
Description
- RFP-Trap Magnetic Particles M-270 is highly recommended, when very large proteins/complexes are investigated, and magnetic separation is needed for IP
- Consists of a RFP VHH/ Nanobody coupled to Magnetic Particles M-270
Specifications
Upon receipt store at 4°C. Do not freeze. | |
mRFP, mCherry, mRFPruby, mPlum, tagRFP, mKate2, mOrange, PA-mCherry, mScarlet | |
Alpaca |
IP, CoIP | |
Magnetic Particles M-270 | |
20 Reactions |