Learn More
Proteintech ChromoTek TurboGFP-Trap Magnetic Agarose
TurboGFP-Trap Magnetic Agarose is an affinity reagent for immunoprecipitation (IP) of TurboGFP fusion proteins.
Brand: Proteintech TBTMA-200
Additional Details : Weight : 0.00240kg
Description
- Comprises an anti-TurboGFP VHH/ Nanobody coupled to magnetic agarose beads
Specifications
Shipped at ambient temperature. Upon receipt store at 4°C. Stable for one year. | |
TurboGFP, CopGFP, Pontellina plumata, maxGFP, maxFP-Green | |
Alpaca |
IP, CoIP, ELISA | |
Magnetic Agarose | |
200 Reactions |