missing translation for 'onlineSavingsMsg'
Learn More

Chrysophenine sodium salt, Thermo Scientific Chemicals

Catalog Number p-7043012
Click to view available options
Quantity:
50 g
100 g
250 g
This item is not returnable. View return policy
2870-32-8
C30H26N4Na2O8S2
680.66
MFCD00007488
YQMJDPHTMKUEHG-RPKDOIGLSA-L
chrysophenine g, chrysophenine j, chrysophenine y, aurophenine o, chrysophenine gp, chrysophenine ns, direct yellow c, benzo yellow lg, erie yellow y, chrysophenine esp
54603156
sodium;5-[(4-ethoxyphenyl)diazenyl]-2-[(E)-2-[4-[(4-ethoxyphenyl)diazenyl]-2-sulfophenyl]ethenyl]benzenesulfonic acid
[Na+].[Na+].CCOC1=CC=C(C=C1)N=NC1=CC=C(\C=C/C2=CC=C(C=C2S([O-])(=O)=O)N=NC2=CC=C(OCC)C=C2)C(=C1)S([O-])(=O)=O
This item is not returnable. View return policy
2870-32-8
C30H26N4Na2O8S2
680.66
MFCD00007488
YQMJDPHTMKUEHG-RPKDOIGLSA-L
chrysophenine g, chrysophenine j, chrysophenine y, aurophenine o, chrysophenine gp, chrysophenine ns, direct yellow c, benzo yellow lg, erie yellow y, chrysophenine esp
54603156
sodium;5-[(4-ethoxyphenyl)diazenyl]-2-[(E)-2-[4-[(4-ethoxyphenyl)diazenyl]-2-sulfophenyl]ethenyl]benzenesulfonic acid
[Na+].[Na+].CCOC1=CC=C(C=C1)N=NC1=CC=C(\C=C/C2=CC=C(C=C2S([O-])(=O)=O)N=NC2=CC=C(OCC)C=C2)C(=C1)S([O-])(=O)=O

A diazo stilbene dye

A diazo stilbene dye

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Orange to Orange-Yellow
Quantity 50 g
Formula Weight 680.66
Physical Form Powder
Chemical Name or Material Chrysophenine sodium salt

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'