missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CHST13 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 20141-1-AP-20UL
Additional Details : Weight : 0.00010kg
Description
The antibody is specific to CHST13.
The protein encoded by this gene belongs to the sulfotransferase 2 family. It is localized to the Golgi membrane, and catalyzes the transfer of sulfate to the C4 hydroxyl of beta-1, 4-linked N-acetylgalactosamine (GalNAc) flanked by glucuronic acid residue in chondroitin. Chondroitin sulfate constitutes the predomit proteoglycan present in cartilage and is distributed on the surfaces of many cells and extracellular matrices.Specifications
CHST13 | |
Polyclonal | |
Unconjugated | |
CHST13 | |
C4ST 3, C4ST3, CHST13 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
166012, 500257, 71797 | |
-20°C | |
Liquid |
Western Blot, Immunohistochemistry (Paraffin), Immunofluorescence, Immunoprecipitation, Immunocytochemistry | |
0.42 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q8NET6 | |
Chst13 | |
Peptide | |
20 μL | |
Primary | |
Mouse, Human, Rat | |
Antibody | |
IgG |