Learn More
CHT1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 21848-1-AP-20UL
Additional Details : Weight : 0.00010kg
Description
Acetylcholine functions as a neurotransmitter in both the central and peripheral nervous systems of all vertebates, and is the principle neurotransmitter used at the neuromuscular junction. Presynaptic synthesis of acetylcholine requires a steady supply of choline, which is acquired by a plasma membrane high affinity choline transporter (CHT).Specifications
CHT1 | |
Polyclonal | |
Unconjugated | |
SLC5A7 | |
CHT, CHT1, hCHT, SLC5A7 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
60482, 63993, 85426 | |
-20°C | |
Liquid |
Immunoprecipitation, Western Blot, Immunohistochemistry (Paraffin) | |
0.46 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q8BGY9, Q9GZV3, Q9JMD7 | |
SLC5A7 | |
CHT1 Fusion Protein Ag16136 | |
20 μL | |
Primary | |
Human, Rat, Mouse | |
Antibody | |
IgG |