missing translation for 'onlineSavingsMsg'
Learn More

CHT1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 21848-1-AP-150UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16881395

  • 3313.85 SEK / 150µL
Estimated Shipment: 07-12-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Acetylcholine functions as a neurotransmitter in both the central and peripheral nervous systems of all vertebates, and is the principle neurotransmitter used at the neuromuscular junction. Presynaptic synthesis of acetylcholine requires a steady supply of choline, which is acquired by a plasma membrane high affinity choline transporter (CHT).
Specifications

Specifications

CHT1
0.46 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q8BGY9, Q9GZV3, Q9JMD7
CHT, CHT1, hCHT, SLC5A7
Rabbit
IgG
150 μL
-20°C
Primary
60482, 63993, 85426
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Unconjugated
SLC5A7
Liquid
SLC5A7
CHT1 Fusion Protein Ag16136
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat
Videos
SDS
Documents
Certificates
Special Offers

Special Offers