Läs mer
Chymase/CMA1/Mast Cell Chymase Antibody [Biotin], Novus Biologicals™
Sheep Polyclonal Antibody
Brand: Novus Biologicals AF4099B
5092.52 SEK valid until 2025-12-16
BÄSTA PRIS på kampanj! Använd kampanjkod: "24090" för att få ditt kampanjpris.
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
Chymase/CMA1/Mast Cell Chymase Polyclonal antibody specifically detects Chymase/CMA1/Mast Cell Chymase in Human samples. It is validated for Immunoprecipitation,Western Blot
Specifikationer
| Chymase/CMA1/Mast Cell Chymase | |
| Polyclonal | |
| PBS | |
| Sheep | |
| Antigen Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Immunoprecipitation, Western Blot | |
| Biotin | |
| Alpha-chymase, chymase, chymase 1 preproprotein transcript E, chymase 1 preproprotein transcript I, chymase 1, mast cell, chymase, heart, chymase, mast cell, CYH, CYM, EC 3.4.21, EC 3.4.21.39, Mast cell protease I, MCT1, MGC119890, MGC119891 | |
| Mouse myeloma cell line NS0-derived recombinant human Chymase/CMA1, Gly20-Asn247, Accession # P23946 | |
| 0.1 mL | |
| Cell Biology, Cellular Markers, Immunology, Innate Immunity, Mast Cell Markers | |
| 1215 | |
| Store at 4°C in the dark. | |
| IgG |