Läs mer
Cibacron Brilliant Yellow 3G-P, Technical Grade, TRC
Produktkod. 30179410
Change view
Kvantitet:
1 g
2.5 g
5 g
Förpackningsstorlek:
2.5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
30179410 5 g -
30179039 1 g -
30179152 2.5 g 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30179410 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC5357335G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 50662-99-2
Molekylformel C25H15Cl3N9Na3O10S3
Molekylvikt (g/mol) 872.96
Synonym 4-[[4-Chloro-6-[(4-sulfophenyl)amino]-1,3,5-triazin-2-yl]amino]-2-[[1-(2,5-dichloro-4-sulfophenyl)-4,5-dihydro-3-methyl-5-oxo-1H-pyrazol-4-yl]azo]-benzenesulfonic Acid trisodium salt, C.I. 18972C.I. Reactive Yellow 2Cibacron Brilliant Yellow 3G-PCibacron Yellow 3GHelaktyn Yellow D 5GNKP Cion Yellow P-VKayacion Yellow P 5GKayacion Yellow P 5G Liquid 20Kayacion Yellow P 5G Liquid 33Ostazin Brilliant Yellow H 5GOstazin Yellow H 5GProcion Brilliant Yellow H 5GProcion Golden Yellow H 5GProcion Yellow H 5GProcion Yellow P 5GNReactive Flavine Yellow K 6GReactive Yellow 2Reactive Yellow K 6GReactive Yellow M 3GPSumifix Brilliant Yellow H 3GXiron Brilliant Yellow 3G-HDYellow P 5G
IUPAC-namn trisodium;2,5-dichloro-4-[4-[[5-[[4-chloro-6-(4-sulfonatoanilino)-1,3,5-triazin-2-yl]amino]-2-sulfonatophenyl]diazenyl]-3-methyl-5-oxo-4H-pyrazol-1-yl]benzenesulfonate
LEDER [Na+].[Na+].[Na+].CC1=NN(C(=O)C1N=Nc2cc(Nc3nc(Cl)nc(Nc4ccc(cc4)S(=O)(=O)[O-])n3)ccc2S(=O)(=O)[O-])c5cc(Cl)c(cc5Cl)S(=O)(=O)[O-]
Specifikationer
Kemiskt namn eller material Cibacron Brilliant Yellow 3G-P, Technical Grade
Kvantitet 5 g
InChI formel InChI=1S/C25H18Cl3N9O10S3.3Na/c1-11-21(22(38)37(36-11)18-9-16(27)20(10-15(18)26)50(45,46)47)35-34-17-8-13(4-7-19(17)49(42,43)44)30-25-32-23(28)31-24(33-25)29-12-2-5-14(6-3-12)48(39,40)41;;;/h2-10,21H,1H3,(H,39,40,41)(H,42,43,44)(H,45,46,47)(H2,29,30,31,32,33);;;/q;3*+1/p-3
Formel vikt 870.8863