Läs mer
Ciclacillin, LoGiCal
Produktkod. 30458053
Change view
Kvantitet:
5 mg
Produktkod. Kvantitet
30458053 5 mg
1 options
Produktkod. 30458053 Leverantör LoGiCal Leverantörsnummer LGCFOR1993.00
LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Ciclacillin
CAS 3485-14-1
Analyt- eller komponentnamn Ciclacillin
Frakt skick Ice pack (-20°C)
Molekylformel C15 H23 N3 O4 S
Certifikat och efterlevnad ISO 9001
Synonym 4-Thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid, 6-[[(1-aminocyclohexyl)carbonyl]amino]-3,3-dimethyl-7-oxo-, (2S,5R,6R)-, 4-Thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid, 6-(1-aminocyclohexanecarboxamido)-3,3-dimethyl-7-oxo- (7CI,8CI), 4-Thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid, 6-[[(1-aminocyclohexyl)carbonyl]amino]-3,3-dimethyl-7-oxo-, [2S-(2α,5α,6β)]-, (1-Aminocyclohexyl)penicillin, 6-(1-Aminocyclohexanecarboxamido)penicillanic acid, 6-(1-Aminocyclohexylcarboxamido)penicillanic acid, Aminocyclohexylpenicillin, Calthor, Ciclacillin, Ciclacillum, Citosarin, Cyclacillin, Cyclapen, NSC 88789, Penicillin, (aminocyclohexyl)-, Syngacillin, Ultracillin, Vastcillin, Vatracin, Vipicil, Wy 4508, Wyvital
InChI formel InChI=1S/C15H23N3O4S/c1-14(2)9(12(20)21)18-10(19)8(11(18)23-14)17-13(22)15(16)6-4-3-5-7-15/h8-9,11H,3-7,16H2,1-2H3,(H,17,22)(H,20,21)/t8-,9+,11-/m1/s1
Lösningstyp Neat
LEDER CC1(C)S[C@@H]2[C@H](NC(=O)C3(N)CCCCC3)C(=O)N2[C@H]1C(=O)O
IUPAC-namn (2S,5R,6R)-6-[(1-aminocyclohexanecarbonyl)amino]-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid
Industry Type Clinical Testing
Molekylvikt (g/mol) 341.43
Kvantitet 5 mg
Formel vikt 341.1409 g/mol