Läs mer
Cicletanine Hydrochloride, TRC
Produktkod. 17980615
Change view
Kvantitet:
1 mg
5 mg
10 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
17980615 10 mg
17931435 5 mg
17953455 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 17980615 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC43270010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 82747-56-6
Molekylformel C14 H12 Cl N O2 . Cl H
Molekylvikt (g/mol) 298.16
Synonym Furo[3,4-c]pyridin-7-ol, 3-(4-chlorophenyl)-1,3-dihydro-6-methyl-, hydrochloride (1:1), Furo[3,4-c]pyridin-7-ol, 3-(4-chlorophenyl)-1,3-dihydro-6-methyl-, hydrochloride (9CI), BN 1270, Cicletanine hydrochloride, Coverine, Justar, Secletan, Tenstaten
IUPAC-namn 3-(4-chlorophenyl)-6-methyl-1,3-dihydrofuro[3,4-c]pyridin-7-ol;hydrochloride
LEDER Cl.Cc1ncc2C(OCc2c1O)c3ccc(Cl)cc3
Specifikationer
Kemiskt namn eller material Cicletanine Hydrochloride
Kvantitet 10 mg
InChI formel InChI=1S/C14H12ClNO2.ClH/c1-8-13(17)12-7-18-14(11(12)6-16-8)9-2-4-10(15)5-3-9;/h2-6,14,17H,7H2,1H3;1H
Formel vikt 297.0323