Läs mer
Tocris Bioscience™ Ciliobrevin A
Beskrivning
Ciliobrevin A is a Hedgehog (Hh) pathway antagonist; blocks Sonic hedgehog (Shh)-induced Hh pathway activation (IC50 = 7 μM) downstream of Smo. Perturbs primary cilia formation; inhibits cytoplasmic AAA+ ATPase dynein-dependent microtubule gliding and ATPase activity.
Specifikationer
Specifikationer
| Kemiskt namn eller material | 2,4-Dichloro-α-(3,4-4-oxo-2(1H)-quinazolinylidene)-β-oxobenzenepropanenitrile |
| CAS | 302803-72-1 |
| Analysprocentintervall | >98% |
| Kvantitet | 25 mg |
| Molekylformel | C17H9Cl2N3O2 |
| Formel vikt | Observed MW: 358.18 |