Läs mer
Cilofungin, TRC
Produktkod. 30353329
Change view
Kvantitet:
2.5 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30353329 10 mg
30351927 2.5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30353329 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC44140010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 79404-91-4
Molekylformel C49H71N7O17
Molekylvikt (g/mol) 1030.12
Synonym 1-[(4R,5R)-4,5-Dihydroxy-N2-[4-(octyloxy)benzoyl]-L-ornithine]-echinocandin B, 1H-Dipyrrolo[2,1-c:2',1'-l][1,4,7,10,13,16]hexaazacycloheneicosine Cyclic Peptide derivative, Antibiotic LY 121019, L 646991, (2alpha,3beta,4beta)-(4R,5R)-4,5-Dihydroxy-N2-[4-(octyloxy)benzoyl]-L-ornithyl-L-threonyl-trans-4-hydroxy-L-prolyl-(S)-4-hydroxy-4-(4-hydroxyphenyl)-L-threonyl-L-threonyl-3-hydroxy-4-methyl-L-proline Cyclic (6-1)-peptide, LY 121019
IUPAC-namn N-[(3S,6S,9S,11R,15S,18S,20R,21R,24S,25S,26S)-6-[(1S,2S)-1,2-dihydroxy-2-(4-hydroxyphenyl)ethyl]-11,20,21,25-tetrahydroxy-3,15-bis[(1R)-1-hydroxyethyl]-26-methyl-2,5,8,14,17,23-hexaoxo-1,4,7,13,16,22-hexazatricyclo[22.3.0.09,13]heptacosan-18-yl]-4-octoxybenzamide
LEDER CCCCCCCCOc1ccc(cc1)C(=O)N[C@H]2C[C@@H](O)[C@@H](O)NC(=O)[C@@H]3[C@@H](O)[C@@H](C)CN3C(=O)[C@@H](NC(=O)[C@@H](NC(=O)[C@@H]4C[C@@H](O)CN4C(=O)[C@@H](NC2=O)[C@@H](C)O)[C@H](O)[C@@H](O)c5ccc(O)cc5)[C@@H](C)O
Specifikationer
Kemiskt namn eller material Cilofungin
Kvantitet 10 mg
InChI formel InChI=1S/C49H71N7O17/c1-5-6-7-8-9-10-19-73-31-17-13-28(14-18-31)42(65)50-32-21-34(61)45(68)54-47(70)38-39(62)24(2)22-56(38)49(72)36(26(4)58)52-46(69)37(41(64)40(63)27-11-15-29(59)16-12-27)53-44(67)33-20-30(60)23-55(33)48(71)35(25(3)57)51-43(32)66/h11-18,24-26,30,32-41,45,57-64,68H,5-10,19-23H2,1-4H3,(H,50,65)(H,51,66)(H,52,69)(H,53,67)(H,54,70)/t24-,25+,26+,30+,32-,33-,34+,35-,36-,37-,38-,39-,40-,41-,45+/m0/s1
Formel vikt 1029.4906