Learn More
Thermo Scientific™ Cimarec™ Extension Cables
Use Thermo Scientific™ Cimarec™ Extension Cables with Cimarec Magnetic Stirrers.
1130.12 SEK - 1775.06 SEK
Product Code | Brand | Description | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Price | Quantity & Availability | |||||
10185411
|
Thermo Scientific™
50088016 |
4-Pin Extension Cable |
1130.12 SEK
Each |
In Stock
Log in to see stock. |
|||||
10013881
|
Thermo Scientific™
50088021 |
6-Pin Extension Cable |
1443.72 SEK
Each |
In Stock
Log in to see stock. |
|||||
10013431
|
Thermo Scientific™
50088102 |
8-Pin Extension Cable |
1775.06 SEK
Each |
Estimated Shipment: 02-07-2024
Log in to see stock. |
|||||
Description
Use Thermo Scientific™ Cimarec™ Extension Cables with Cimarec Magnetic Stirrers.
- Length: 3m (9.8′)
Specifications
3 m | |
Extension Cable |