Läs mer
Cimaterol, TRC
Produktkod. 30134910
Change view
Kvantitet:
10 mg
25 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30134910 25 mg
30134907 50 mg
30006584 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30134910 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC44160025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 54239-37-1
Molekylformel C12 H17 N3 O
Molekylvikt (g/mol) 219.28
Synonym 2-Amino-5-[1-hydroxy-2-[(1-methylethyl)amino]ethyl]benzonitrile, CL 263780, Cimaterol
IUPAC-namn 2-amino-5-[1-hydroxy-2-(propan-2-ylamino)ethyl]benzonitrile
LEDER CC(C)NCC(O)c1ccc(N)c(c1)C#N
Specifikationer
Kemiskt namn eller material Cimaterol
Anteckningar om renhetsgrad HPLC
Kvantitet 25 mg
InChI formel InChI=1S/C12H17N3O/c1-8(2)15-7-12(16)9-3-4-11(14)10(5-9)6-13/h3-5,8,12,15-16H,7,14H2,1-2H3
Formel vikt 219.1372
Procent renhet >95%