Läs mer
Cimbuterol, TRC
Produktkod. 30009124
Change view
Kvantitet:
10 mg
25 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30009124 100 mg
30003814 25 mg
30008784 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30009124 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC441620100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 54239-39-3
Molekylformel C13 H19 N3 O
Molekylvikt (g/mol) 233.31
Synonym Benzonitrile, 2-amino-5-[2-[(1,1-dimethylethyl)amino]-1-hydroxyethyl]-, 2-Amino-5-[2-[(1,1-dimethylethyl)amino]-1-hydroxyethyl]benzonitrile, Cimbuterol
IUPAC-namn 2-amino-5-[2-(tert-butylamino)-1-hydroxyethyl]benzonitrile
LEDER CC(C)(C)NCC(O)c1ccc(N)c(c1)C#N
Specifikationer
Kemiskt namn eller material Cimbuterol
Anteckningar om renhetsgrad HPLC
Kvantitet 100 mg
InChI formel InChI=1S/C13H19N3O/c1-13(2,3)16-8-12(17)9-4-5-11(15)10(6-9)7-14/h4-6,12,16-17H,8,15H2,1-3H3
Formel vikt 233.1528
Procent renhet >95%