Läs mer
Cimetidine EP Impurity A, Mikromol™
Produktkod. 30050372
Change view
Kvantitet:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30050372 100 mg
30050658 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30050372 Leverantör Mikromol Leverantörsnummer MM0020.08
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
Cimetidine EP Impurity A
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 3-Cyano-2-methyl-1-[2-[((5-methyl-1H-imidazol-4-yl)methyl)sulphanyl]ethyl]isothiourea
CAS 52378-40-2
Analyt- eller komponentnamn 3-Cyano-2-methyl-1-[2-[((5-methyl-1H-imidazol-4-yl)methyl)sulphanyl]ethyl]isothiourea
Frakt skick Room Temperature
Molekylformel C10 H15 N5 S2
Certifikat och efterlevnad ISO 17025
Synonym 3-Cyano-2-methyl-1-[2-[[(5-methyl-1H-imidazol-4-yl)methyl]sulphanyl]ethyl]isothiourea, Cimetidine Hydrochloride Imp. A (EP), Cimetidine Imp. A (EP), 3-Cyano-2-methyl-1-[2-[[(5-methyl-1H-imidazol-4-yl)methyl]sulphanyl]ethyl]isothiourea, Cimetidine Hydrochloride Impurity A, Cimetidine Impurity A, Cimetidine Impurity A
InChI formel InChI=1S/C10H15N5S2/c1-8-9(15-7-14-8)5-17-4-3-12-10(16-2)13-6-11/h7H,3-5H2,1-2H3,(H,12,13)(H,14,15)
LEDER CSC(=NC#N)NCCSCc1nc[nH]c1C
IUPAC-namn methyl N'-cyano-N-[2-[(5-methyl-1H-imidazol-4-yl)methylsulfanyl]ethyl]carbamimidothioate
Molekylvikt (g/mol) 269.39
Kvantitet 100 mg
Formel vikt 269.0769 g/mol