Läs mer
Cimetidine EP Impurity H, Mikromol™
Produktkod. 30460983
Change view
Kvantitet:
25 mg
100 mg
4 x 25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30460983 4 x 25 mg
17939609 100 mg
18030800 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30460983 Leverantör Mikromol Leverantörsnummer MM0020.0700254AMP
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
Cimetidine EP Impurity H, Mikromol
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 1,1'-(Disulfanediyldiethylene)bis(2-cyano-3-methylguanidine)
CAS 74886-59-2
Analyt- eller komponentnamn 1,1'-(Disulfanediyldiethylene)bis(2-cyano-3-methylguanidine)
Frakt skick Room Temperature
Molekylformel C10 H18 N8 S2
Certifikat och efterlevnad ISO 17025
Synonym 1,1'-(Disulphanediyldiethylene)bis(2-cyano-3-methylguanidine), Cimetidine Hydrochloride Imp. H (EP), Cimetidine Imp. H (EP), 1,1'-(Disulphanediyldiethylene)bis(2-cyano-3-methylguanidine), Cimetidine Hydrochloride Impurity H, Cimetidine Impurity H, Cimetidine Impurity H
InChI formel InChI=1S/C10H18N8S2/c1-13-9(17-7-11)15-3-5-19-20-6-4-16-10(14-2)18-8-12/h3-6H2,1-2H3,(H2,13,15,17)(H2,14,16,18)
Lösningstyp Neat
LEDER CNC(=NC#N)NCCSSCCNC(=NC#N)NC
IUPAC-namn 2-cyano-1-[2-[2-[(N'-cyano-N-methylcarbamimidoyl)amino]ethyldisulfanyl]ethyl]-3-methylguanidine
Molekylvikt (g/mol) 314.43
Kvantitet 4 x 25 mg
Formel vikt 314.1096 g/mol