Läs mer
Cimetropium Bromide, TRC
Produktkod. 30356233
Change view
Kvantitet:
1 g
Produktkod. Kvantitet
30356233 1 g
1 options
Produktkod. 30356233 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC4416851G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Cimetropium Bromide
CAS 51598-60-8
Molekylformel C21 H28 N O4 . Br
Kvantitet 1 g
Synonym 3-Oxa-9-azoniatricyclo[3.3.1.02,4]nonane, 9-(cyclopropylmethyl)-7-[(2S)-3-hydroxy-1-oxo-2-phenylpropoxy]-9-methyl-, bromide (1:1), (1α,2β,4β,5α,7β)-, 3-Oxa-9-azoniatricyclo[3.3.1.02,4]nonane, 9-(cyclopropylmethyl)-7-(3-hydroxy-1-oxo-2-phenylpropoxy)-9-methyl-, bromide, [7(S)-(1α,2β,4β,5α,7β)]-, 3-Oxa-9-azoniatricyclo[3.3.1.02,4]nonane, 9-(cyclopropylmethyl)-7-[(2S)-3-hydroxy-1-oxo-2-phenylpropoxy]-9-methyl-, bromide, (1α,2β,4β,5α,7β)- (9CI), 3-Oxa-9-azatricyclo[3.3.1.02,4]nonane, 3-oxa-9-azoniatricyclo[3.3.1.02,4]nonane deriv., Alginor, Cimetropium bromide, DA 3177, N-Cyclopropylmethylscopolamine bromide
InChI formel InChI=1S/C21H28NO4.BrH/c1-22(11-13-7-8-13)17-9-15(10-18(22)20-19(17)26-20)25-21(24)16(12-23)14-5-3-2-4-6-14;/h2-6,13,15-20,23H,7-12H2,1H3;1H/q+1;/p-1/t15-,16-,17-,18+,19-,20+,22?;/m1./s1
LEDER [Br-].C[N+]1(CC2CC2)[C@@H]3C[C@H](C[C@H]1[C@@H]4O[C@H]34)OC(=O)[C@H](CO)c5ccccc5
IUPAC-namn [(1R,2R,4S,5S)-9-(cyclopropylmethyl)-9-methyl-3-oxa-9-azoniatricyclo[3.3.1.02,4]nonan-7-yl] (2S)-3-hydroxy-2-phenylpropanoate;bromide
Molekylvikt (g/mol) 438.36
Formel vikt 437.1202