Läs mer
Cinaciguat hydrochloride, MedChemExpress
Produktkod. 16490206
Encompass_Preferred
Change view
Kvantitet:
2 mg
5 mg
10 mg
50 mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
16490206 2 mg
16472127 50 mg
16428607 5 mg
16425988 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Produktkod. 16490206 Leverantör Medchem Express Leverantörsnummer HY14181A
MedChemExpress Cinaciguat hydrochloride is a potent soluble guanylate cyclase (GC) activator with EC50 of 15 nM in platelets.
Cinaciguat hydrochloride is a potent soluble guanylate cyclase (GC) activator with EC50 of 15 nM in platelets.
Specifikationer
Kemiskt namn eller material Cinaciguat hydrochloride
CAS 646995-35-9
Färg White
Anteckningar om renhetsgrad Research
Molekylformel C36H40ClNO5
Kvantitet 2 mg
Synonym BAY 58-2667 hydrochloride
Löslighetsinformation DMSO : 110 mg/mL (182.68 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (ultrasonic;warming;heat to 60°C) (insoluble)
LEDER OC(C(C=C1)=CC=C1CN(CCCCC(O)=O)CCC(C=CC=C2)=C2OCC(C=C3)=CC=C3CCC4=CC=CC=C4)=O.[H]Cl
Molekylvikt (g/mol) 602.16
Formel vikt 602.16
Procent renhet 99.52%
Kvalitet Research
Fysisk form Solid
För användning med (applikation) COVID-19-immunoregulation