missing translation for 'onlineSavingsMsg'
Learn More

Cineole, Approx. 99%, MP Biomedicals™

Catalog Number p-4679708
Click to view available options
Quantity:
50 mL
500 mL
This item is not returnable. View return policy
470-82-6
C10H18O
154.25
MFCD00167977
WEEGYLXZBRQIMU-UHFFFAOYSA-N
eucalyptol, cineole, 1,8-cineole, 1,8-cineol, cajeputol, 1,8-epoxy-p-menthane, eucalyptole, zineol, eucapur, terpan
2758
1,3,3-trimethyl-2-oxabicyclo[2.2.2]octane
CC12CCC(CC1)C(C)(C)O2
This item is not returnable. View return policy
470-82-6
C10H18O
154.25
MFCD00167977
WEEGYLXZBRQIMU-UHFFFAOYSA-N
eucalyptol, cineole, 1,8-cineole, 1,8-cineol, cajeputol, 1,8-epoxy-p-menthane, eucalyptole, zineol, eucapur, terpan
2758
1,3,3-trimethyl-2-oxabicyclo[2.2.2]octane
CC12CCC(CC1)C(C)(C)O2
Pharmaceuticals, Insecticide
Color Colorless
Density 0.921 to 0.924g/mL
Assay Percent Range Approx. 99%
Quantity 50 mL
Formula Weight 154.3
Physical Form Liquid
Chemical Name or Material Cineole
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'