Läs mer
Cinnamylamine Hydrochloride, TRC
Produktkod. 30349212
Change view
Kvantitet:
100 mg
500 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30349212 100 mg
30351080 500 mg
30368309 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30349212 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC442090100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 5586-89-0
Molekylformel C9 H11 N . Cl H
Molekylvikt (g/mol) 169.65
Synonym 3-Phenyl-2-Propen-1-amine Hydrochloride (1:1), 2-Propen-1-amine, 3-phenyl-, hydrochloride (9CI), Cinnamylamine, hydrochloride (6CI,7CI,8CI)
IUPAC-namn (E)-3-phenylprop-2-en-1-amine;hydrochloride
LEDER Cl.NC\C=C\c1ccccc1
Specifikationer
Kemiskt namn eller material Cinnamylamine Hydrochloride
Kvantitet 100 mg
InChI formel InChI=1S/C9H11N.ClH/c10-8-4-7-9-5-2-1-3-6-9;/h1-7H,8,10H2;1H/b7-4+;
Formel vikt 169.0658