missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ CIRBP (Human) Recombinant Protein
Human CIRBP full-length ORF ( AAH00403, 1 a.a. - 172 a.a.) recombinant protein with GST-tag at N-terminal.
Brand: Abnova™ H00001153-P02.10ug
Additional Details : Weight : 0.00010kg
Description
- Sequence: MASDEGKLFVGGLSFDTNEQSLEQVFSKYGQISEVVVVKDRETQRSRGFGFVTFENIDDAKDAMMAMNGKSVDGRQIRVDQAGKSSDNRSRGYRGGSAGGRGFFRGGRGRGRGFSRGGGDRGYGGNRFESRSGGYGGSRDYYSSRSQSGGYSDRSSGGSYRDSYDSYATHNE
Specifications
AAH00403 | |
cold inducible RNA binding protein | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
CIRBP | |
GST |
1153 | |
Wheat germ expression system | |
10 μg | |
CIRP | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |