Läs mer
Heinz Herenz Circular Cover Glasses
Produktkod. 30158500
Change view
Bredd (metrisk):
10 mm
11 mm
12 mm
13 mm
14 mm
15 mm
18 mm
20 mm
22 mm
24 mm
25 mm
30 mm
32 mm
13 product options available for selection
Product selection table with 13 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Bredd (metrisk)
30158500 25 mm
30158480 10 mm
30158326 14 mm
30158414 24 mm
30158437 13 mm
30158495 20 mm
30158325 12 mm
30158473 18 mm
30158440 22 mm
30158374 11 mm
30158498 15 mm
30158344 30 mm
30158420 32 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 13 options available.
13 options
Produktkod. 30158500 Leverantör Heinz Herenz Leverantörsnummer 1051209
Bara null kvar
Lägg i varukorgen
Lägg i varukorgen
Circular cover glasses.
  • Round cover glasses
Specifikationer
Produkttyp Coverslip
Form Circle
Material Borosilicate Glass
Längd (metrisk) 24 mm
Bredd (metrisk) 25 mm
Tjocklek (metrisk) 1 mm
Sterilitet Non-sterile
Disponibel Single-use