missing translation for 'onlineSavingsMsg'
Läs mer

cis-1,2,3,4-Tetrahydro-1-phenyl-4-isoquinolinol, TRC

Produktkod. 30416774
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30416774 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 30416774 Leverantör Toronto Research Chemicals Leverantörsnummer TRCT2942955MG

Please to purchase this item. Need a web account? Register with us today!

Denna artikel kan inte returneras. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifikationer

Kemiskt namn eller material cis-1,2,3,4-Tetrahydro-1-phenyl-4-isoquinolinol
CAS 90861-85-1
Molekylformel C15H15NO
Kvantitet 5 mg
Synonym (1R,4S)-rel-1,2,3,4-Tetrahydro-1-phenyl-4-isoquinolinol
InChI formel InChI=1S/C15H15NO/c17-14-10-16-15(11-6-2-1-3-7-11)13-9-5-4-8-12(13)14/h1-9,14-17H,10H2/t14-,15-/m0/s1
LEDER O[C@H]1CN[C@@H](c2ccccc2)c3ccccc13
IUPAC-namn (1S,4R)-1-phenyl-1,2,3,4-tetrahydroisoquinolin-4-ol
Molekylvikt (g/mol) 225.29
Formel vikt 225.1154
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.