Läs mer
cis-1,3-Dibenzyl-2-imidazolidone-4,5-dicarboxylic Acid, TRC
Produktkod. 30377300
Change view
Kvantitet:
25 g
Produktkod. Kvantitet
30377300 25 g
1 options
Produktkod. 30377300 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD42268525G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material cis-1,3-Dibenzyl-2-imidazolidone-4,5-dicarboxylic Acid
CAS 51591-75-4
Molekylformel C19H18N2O5
Kvantitet 25 g
Synonym cis-1,3-Dibenzyl-2-oxo-4,5-imidazolidinedicarboxylic Acid, cis-2-oxo-1,3-Bis(phenylmethyl)-4,5-Imidazolidinedicarboxylic Acid, (4R,5S)-rel-2-Oxo-1,3-Bis(phenylmethyl)-4,5-imidazolidinedicarboxylic Acid
InChI formel InChI=1S/C19H18N2O5/c22-17(23)15-16(18(24)25)21(12-14-9-5-2-6-10-14)19(26)20(15)11-13-7-3-1-4-8-13/h1-10,15-16H,11-12H2,(H,22,23)(H,24,25)/t15-,16+
LEDER OC(=O)[C@@H]1[C@@H](N(Cc2ccccc2)C(=O)N1Cc3ccccc3)C(=O)O
Molekylvikt (g/mol) 354.36