Läs mer
cis-2-(2,4-Dichlorophenyl)-2-(1H-imidazol-1-ylmethyl)-1,3-dioxolane-4-methanol, TRC
Produktkod. 30009691
Change view
Kvantitet:
250 mg
2.5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30009691 2.5 g
30003836 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30009691 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD4359302.5G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 61397-58-8
Molekylformel C14 H14 Cl2 N2 O3
Molekylvikt (g/mol) 329.18
Synonym (2R,4S)-rel-2-(2,4-Dichlorophenyl)-2-(1H-imidazol-1-ylmethyl)-1,3-dioxolane-4-methanol, 1,3-Dioxolane-4-methanol, 2-(2,4-dichlorophenyl)-2-(1H-imidazol-1-ylmethyl)-, cis-, rel-(2R,4S)-2-(2,4-Dichlorophenyl)-2-(1H-imidazol-1-ylmethyl)-1,3-dioxolane-4-methanol, [(2RS,4SR)-2-(2,4-Dichlorophenyl)-2-(1-Himidazol-1-ylmethyl)-1,3-dioxolan-4-yl]methanol
IUPAC-namn ((2RS,4SR)-2-(2,4-Dichlorophenyl)-2-(1-Himidazol-1-ylmethyl)-1,3-dioxolan-4-yl)methanol
LEDER OC[C@H]1CO[C@@](Cn2ccnc2)(O1)c3ccc(Cl)cc3Cl
Specifikationer
Kemiskt namn eller material ((2RS,4SR)-2-(2,4-Dichlorophenyl)-2-(1-Himidazol-1-ylmethyl)-1,3-dioxolan-4-yl)methanol
Kvantitet 2.5 g
Anteckningar om renhetsgrad HPLC
InChI formel InChI=1S/C14H14Cl2N2O3/c15-10-1-2-12(13(16)5-10)14(8-18-4-3-17-9-18)20-7-11(6-19)21-14/h1-5,9,11,19H,6-8H2/t11-,14-/m0/s1
Formel vikt 328.0381
Procent renhet >95%