Learn More
cis-2-Penten-1-ol, 97%, remainder mainly trans-isomer
Product Code. 11422407
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11422407 5 g 5g
11432407 25 g 25g
11442407 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 11422407 Supplier Thermo Scientific Alfa Aesar Supplier No. A14262.06
 Restricted Item
Beräknad leverans: 27-11-2026
Add to basket
CAS: 1576-95-0 | C5H10O | 86.13 g/mol
cis-2-Penten-1-ol is used in the preparation of 1-bromo-pent-2-ene by reacting with phosphorus tribromide and pyridine. Further, it is a banana flavored compound.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
cis-2-Penten-1-ol is used in the preparation of 1-bromo-pent-2-ene by reacting with phosphorus tribromide and pyridine. Further, it is a banana flavored compound.
Solubility
Miscible with alcohol. Immiscible with water.
Notes
Incompatible with strong oxidizing agents, acid chlorides and acid anhydrides.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 1576-95-0
Molecular Formula C5H10O
Molecular Weight (g/mol) 86.13
MDL Number MFCD00063208
InChI Key BTSIZIIPFNVMHF-ARJAWSKDSA-N
Synonym cis-2-penten-1-ol, z-pent-2-en-1-ol, z-2-penten-1-ol, 2-penten-1-ol, z, e-2-penten-1-ol, unii-qnc2nb53mj, 2z-penten-1-ol, cis-2-pentenol, qnc2nb53mj, pent-2 e-enol
PubChem CID 5364919
SMILES CC\C=C/CO
Specifications
Density 0.853
Boiling Point 138°C
Flash Point 48°C (118°F)
Refractive Index 1.436
Quantity 5 g
UN Number UN1987
Beilstein 1719473
Solubility Information Miscible with alcohol. Immiscible with water.
IUPAC Name (2Z)-pent-2-en-1-ol
Formula Weight 86.13
Percent Purity 97%
Assay remainder mainly trans-isomer
Chemical Name or Material cis-2-Penten-1-ol