Läs mer
cis-4-(1,2,3,4-Tetrahydro-6-methoxy-2-phenyl-1-naphthalenyl)phenol, TRC
Produktkod. 30423189
Change view
Kvantitet:
25 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30423189 25 mg
30422762 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30423189 Leverantör Toronto Research Chemicals Leverantörsnummer TRCT29386525MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material cis-4-(1,2,3,4-Tetrahydro-6-methoxy-2-phenyl-1-naphthalenyl)phenol
CAS 14089-22-6
Molekylformel C23H22O2
Kvantitet 25 mg
Synonym cis-4-(6-Methoxy-2-phenyl-1,2,3,4-tetrahydro-1-naphthalenyl)phenol, rel-4-[(1R,2S)-1,2,3,4-Tetrahydro-6-methoxy-2-phenyl-1-naphthalenyl]phenol
InChI formel InChI=1S/C23H22O2/c1-25-20-12-14-22-18(15-20)9-13-21(16-5-3-2-4-6-16)23(22)17-7-10-19(24)11-8-17/h2-8,10-12,14-15,21,23-24H,9,13H2,1H3/t21-,23+/m1/s1
LEDER COC1=CC2=C(C=C1)[C@H]([C@H](CC2)C3=CC=CC=C3)C4=CC=C(C=C4)O
IUPAC-namn 4-[(1R,2S)-6-methoxy-2-phenyl-1,2,3,4-tetrahydronaphthalen-1-yl]phenol
Molekylvikt (g/mol) 330.42
Formel vikt 330.162