missing translation for 'onlineSavingsMsg'
Learn More
Learn More
cis-4-(Boc-amino)cyclohexylamine, 97%, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
25 g
Description
cis-4-(Boc-amino)cyclohexylamine can be used in agrochemical, pharmaceutical and dyestuff field.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
cis-4-(Boc-amino)cyclohexylamine can be used in agrochemical, pharmaceutical and dyestuff field.
Solubility
Soluble in methanol, ethyl acetate and Slightly soluble in water.
Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
cis-4-(Boc-amino)cyclohexylamine can be used in agrochemical, pharmaceutical and dyestuff field.
Solubility
Soluble in methanol, ethyl acetate and Slightly soluble in water.
Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
Specifications
Quantity | 1 g |
Solubility Information | Soluble in methanol,ethyl acetate and Slightly soluble in water. |
Formula Weight | 214.3 |
Percent Purity | 97% |
Chemical Name or Material | cis-4-(Boc-amino)cyclohexylamine |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'