Läs mer
cis-5-(3,5-Dimethoxystyryl)-2-methoxyphenol, Thermo Scientific™
Produktkod. 15476959
Change view
Kvantitet:
10 mg
Förpackningsstorlek:
10mg
Produktkod. Kvantitet unitSize
15476959 10 mg 10mg
1 options
Produktkod. 15476959 Leverantör Thermo Scientific Alfa Aesar Leverantörsnummer J65540.MA
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternativa produkter
A natural polyphenolic antioxidant
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material cis-5-(3,5-Dimethoxystyryl)-2-methoxyphenol
CAS 586410-08-4
Färg Yellow
Molekylformel C17H18O4
MDL-nummer MFCD04973652
Kvantitet 10 mg
Synonym cis-3,4',5-trimethoxy-3'-hydroxystilbene, 5-1z-2-3,5-dimethoxyphenyl ethenyl-2-methoxy-phenol, z-3,4',5-trimethoxy-3'-hydroxystilbene, cis-3,5,4'-trimethoxy-3'-hydroxy stilbene, cis-3,4/',5-trimethoxy-3/'-hydroxystilbene, cis-5-3,5-dimethoxystyryl-2-methoxyphenol, 5-z-2-3,5-dimethoxyphenyl ethenyl-2-methoxyphenol
InChI-nyckel UQIWTPQGJCCTPA-PLNGDYQASA-N
LEDER COC1=C(C=C(C=C1)C=CC2=CC(=CC(=C2)OC)OC)O
IUPAC-namn 5-[(Z)-2-(3,5-dimethoxyphenyl)ethenyl]-2-methoxyphenol
Molekylvikt (g/mol) 286.327
PubChem CID 10108048
Formel vikt 286.32
Fysisk form Oil