missing translation for 'onlineSavingsMsg'
Learn More

cis-Bis(2,2'-bipyridine)dichlororuthenium(II) hydrate, 97%, Thermo Scientific Chemicals

Catalog Number p-3593404
Click to view available options
Quantity:
250 mg
500 mg
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
98014-14-3
C20H16Cl2N4Ru
484.35
MFCD00150005
MGAJEYXQYJBLQL-UHFFFAOYSA-L
cis-bis 2,2'-bipyridine dichlororuthenium ii hydrate, bis bipyridyl dichlororuthenium hydrate, 2,2'-bipyridine-dichlororuthenium-water 2/1/1, cis-dichlorobis 2,2'-bipyridine ruthenium ii, cis-bis 2,2'-bipyridine dichlororuthenium ii monohydrate, cis-bis 2,2-bipyridine dichlororuthenium ii hydrate, cis-bis-2,2'-bipyridine ruthenium ii chloride,hydrate, cis-bis 2,2 inverted exclamation marka-bipyridine dichlororuthenium ii hydrate
16211844
Cl[Ru]Cl.C1=CN=C(C=C1)C1=NC=CC=C1.C1=CN=C(C=C1)C1=NC=CC=C1
This item is not returnable. View return policy
98014-14-3
C20H16Cl2N4Ru
484.35
MFCD00150005
MGAJEYXQYJBLQL-UHFFFAOYSA-L
cis-bis 2,2'-bipyridine dichlororuthenium ii hydrate, bis bipyridyl dichlororuthenium hydrate, 2,2'-bipyridine-dichlororuthenium-water 2/1/1, cis-dichlorobis 2,2'-bipyridine ruthenium ii, cis-bis 2,2'-bipyridine dichlororuthenium ii monohydrate, cis-bis 2,2-bipyridine dichlororuthenium ii hydrate, cis-bis-2,2'-bipyridine ruthenium ii chloride,hydrate, cis-bis 2,2 inverted exclamation marka-bipyridine dichlororuthenium ii hydrate
16211844
Cl[Ru]Cl.C1=CN=C(C=C1)C1=NC=CC=C1.C1=CN=C(C=C1)C1=NC=CC=C1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Black to Green
Infrared Spectrum Authentic
Assay Percent Range 96% min. (Based on Carbon)
Packaging Glass bottle
Quantity 250 mg
Solubility Information Solubility in water: soluble
IUPAC Name dichlororuthenium;2-pyridin-2-ylpyridine;hydrate
Formula Weight 484.35
Percent Purity 97%
Physical Form Powder
Chemical Name or Material cis-Bis(2, 2'-bipyridine)dichlororuthenium(II) hydrate
Show More Show Less
compliance-icons
Product Identifier
  • cis-Bis(2,2'-bipyridine)dichlororuthenium(II) hydrate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'