Läs mer
cis-Clomiphene Citrate, TRC
Produktkod. 30005931
Change view
Kvantitet:
5 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30005931 50 mg
30005155 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30005931 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC58702650MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 7619-53-6
Molekylformel C26 H28 Cl N O . C6 H8 O7
Molekylvikt (g/mol) 598.08
Synonym Ethanamine, 2-[4-[(1Z)-2-chloro-1,2-diphenylethenyl]phenoxy]-N,N-diethyl-, 2-hydroxy-1,2,3-propanetricarboxylate (1:1), Ethanamine, 2-[4-(2-chloro-1,2-diphenylethenyl)phenoxy]-N,N-diethyl-, (Z)-, 2-hydroxy-1,2,3-propanetricarboxylate (1:1), Triethylamine, 2-[p-(2-chloro-1,2-diphenylvinyl)phenoxy]-, citrate (1:1), (Z)- (8CI), (Z)-Clomiphene citrate, Clomiphene A citrate, NSC 151466, Zuclomid, Zuclomiphene citrate, cis-Clomiphene citrate, Zuclomifene Citrate
IUPAC-namn 2-[4-[(Z)-2-chloro-1,2-diphenylethenyl]phenoxy]-N,N-diethylethanamine;2-hydroxypropane-1,2,3-tricarboxylic acid
LEDER CCN(CC)CCOc1ccc(cc1)\C(=C(/Cl)\c2ccccc2)\c3ccccc3.OC(=O)CC(O)(CC(=O)O)C(=O)O
Specifikationer
Kemiskt namn eller material Zuclomifene Citrate
Anteckningar om renhetsgrad HPLC
Kvantitet 50 mg
InChI formel InChI=1S/C26H28ClNO.C6H8O7/c1-3-28(4-2)19-20-29-24-17-15-22(16-18-24)25(21-11-7-5-8-12-21)26(27)23-13-9-6-10-14-23;7-3(8)1-6(13,5(11)12)2-4(9)10/h5-18H,3-4,19-20H2,1-2H3;13H,1-2H2,(H,7,8)(H,9,10)(H,11,12)/b26-25-;
Formel vikt 597.2129
Procent renhet >95%