Läs mer
cis-Cyclohexane-1,2-dimethanol Dimethanesulfonate, TRC
Produktkod. 30361831
Change view
Kvantitet:
50 mg
250 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30361831 50 mg
30360575 250 mg
30356345 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30361831 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC54642050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 66347-68-0
Molekylformel C10H20O6S2
Molekylvikt (g/mol) 300.38
Synonym (1R,2S)-Cyclohexane-1,2-diylbis(methylene) Dimethanesulfonate, NSC 170782, (1R,2S)-rel-1,2-Cyclohexanedimethanol Dimethanesulfonate
IUPAC-namn [2-(methylsulfonyloxymethyl)cyclohexyl]methyl methanesulfonate
LEDER CS(=O)(=O)OC[C@@H]1CCCC[C@@H]1COS(=O)(=O)C
Specifikationer
Kemiskt namn eller material cis-Cyclohexane-1,2-dimethanol Dimethanesulfonate
Kvantitet 50 mg
InChI formel InChI=1S/C10H20O6S2/c1-17(11,12)15-7-9-5-3-4-6-10(9)8-16-18(2,13)14/h9-10H,3-8H2,1-2H3
Formel vikt 300.0701