Läs mer
cis-Diammine-diiodo Platinum II, TRC
Produktkod. 30354977
Change view
Kvantitet:
100 mg
250 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30354977 250 mg
30361400 500 mg
30360388 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30354977 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD416260250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 13841-96-8
Molekylformel H6I2N2Pt
Molekylvikt (g/mol) 482.95
Synonym Diamminediiodo-platinum, Diamminediiodoplatinum
LEDER I[Pt-2]([NH3+])([NH3+])I
Specifikationer
Kemiskt namn eller material cis-Diammine-diiodo Platinum II