Läs mer
cis-Linalool Oxide, TRC
Produktkod. 30469394
Change view
Kvantitet:
5 mg
Produktkod. Kvantitet
30469394 5 mg
1 options
Produktkod. 30469394 Leverantör Toronto Research Chemicals Leverantörsnummer TRCL4659705MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material cis-Linalool Oxide
CAS 5989-33-3
Anteckningar om renhetsgrad GC
Molekylformel C10H18O2
Kvantitet 5 mg
Synonym cis-5-Ethenyltetrahydro-α,α,5-trimethyl-2-furanmethanol, Tetrahydro-α,α,5-trimethyl-5-vinyl-furfuryl Alcohol, rel-(2R,5S)-5-Ethenyltetrahydro-α,α,5-trimethyl-2-furanmethanol, (Z)-Furanoid Linalool Oxide, (Z)-Linalool Oxide B, Epoxydihydrolinalool II, Linalool B Oxide, Linalool Oxide, Linalool Oxide 1, Linalool Oxide B, Linalool Oxide I, cis-2-Methyl-2-vinyl-5-(1-hydroxy-1-methylethyl)tetrahydrofuran, cis-2-Vinyl-2-methyl-5-(1-hydroxy-1-methylethyl)tetrahydrofuran, cis-Furan Linalool Oxide, cis-Furanoid Linalool Oxide, cis-Linalool 3,6-Oxide, cis-Linalool Oxide (furan), cis-Linalool Oxide (furanoid), cis-f-Linalool Oxide, cis-α,α,5-Trimethyl-5-vinyltetrahydrofurfuryl Alcohol
InChI formel InChI=1S/C10H18O2/c1-5-10(4)7-6-8(12-10)9(2,3)11/h5,8,11H,1,6-7H2,2-4H3/t8-,10-/m1/s1
LEDER C=C[C@]1(O[C@@H](C(C)(O)C)CC1)C
IUPAC-namn 2-[(2R,5S)-5-ethenyl-5-methyloxolan-2-yl]propan-2-ol
Molekylvikt (g/mol) 170.25
Formel vikt 170.1307