Läs mer
cis-Montelukast, TRC
Produktkod. 30138305
Change view
Kvantitet:
1 mg
5 mg
10 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30138305 5 mg
17904445 1 mg
17963255 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30138305 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM5680055MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 774538-96-4
Molekylformel C35H36ClNO3S
Molekylvikt (g/mol) 586.18
Synonym Cyclopropaneacetic acid, 1-[[[(1R)-1-[3-[(1Z)-2-(7-chloro-2-quinolinyl)ethenyl]phenyl]-3-[2-(1-hydroxy-1-methylethyl)phenyl]propyl]thio]methyl]-, 1-[[[(1R)-1-[3-[(1Z)-2-(7-Chloro-2-quinolinyl)ethenyl]phenyl]-3-[2-(1-hydroxy-1-methylethyl)phenyl]propyl]thio]methyl]-cyclopropaneacetic acid, Montelukast Sodium Imp. G (EP), cis-Montelukast, (Z)-Montelukast
IUPAC-namn 2-[1-[[(1R)-1-[3-[(Z)-2-(7-chloroquinolin-2-yl)ethenyl]phenyl]-3-[2-(2-hydroxypropan-2-yl)phenyl]propyl]sulfanylmethyl]cyclopropyl]acetic acid
LEDER CC(C)(O)c1ccccc1CC[C@@H](SCC2(CC(=O)O)CC2)c3cccc(\C=C/c4ccc5ccc(Cl)cc5n4)c3
Specifikationer
Kemiskt namn eller material cis-Montelukast
Anteckningar om renhetsgrad HPLC
Kvantitet 5 mg
InChI formel InChI=1S/C35H36ClNO3S/c1-34(2,40)30-9-4-3-7-25(30)13-17-32(41-23-35(18-19-35)22-33(38)39)27-8-5-6-24(20-27)10-15-29-16-12-26-11-14-28(36)21-31(26)37-29/h3-12,14-16,20-21,32,40H,13,17-19,22-23H2,1-2H3,(H,38,39)/b15-10-/t32-/m1/s1
Formel vikt 585.2104
Procent renhet >95%