Läs mer
cis-Octahydroisoindole, TRC
Produktkod. 30418536
Change view
Kvantitet:
500 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30418536 1 g
30410026 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30418536 Leverantör Toronto Research Chemicals Leverantörsnummer TRCO2369451G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material cis-Octahydroisoindole
CAS 1470-99-1
Molekylformel C8H15N
Kvantitet 1 g
Synonym (3aR,7aS)-rel-Octahydro-1H-isoindole, cis-Hexahydroisoindoline, cis-Octahydro-1H-isoindole
InChI formel InChI=1S/C8H15N/c1-2-4-8-6-9-5-7(8)3-1/h7-9H,1-6H2/t7-,8+
LEDER C1CC[C@H]2CNC[C@H]2C1
IUPAC-namn (3aR,7aS)-2,3,3a,4,5,6,7,7a-octahydro-1H-isoindole
Molekylvikt (g/mol) 125.21
Formel vikt 125.1204