Läs mer
cis-Resveratrol, TRC
Produktkod. 17994465
Change view
Kvantitet:
5 mg
10 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
17994465 5 mg
17941605 10 mg
17914195 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 17994465 Leverantör Toronto Research Chemicals Leverantörsnummer TRCR1500055MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 61434-67-1
Molekylformel C14 H12 O3
Molekylvikt (g/mol) 228.24
Synonym 1,3-Benzenediol, 5-[(1Z)-2-(4-hydroxyphenyl)ethenyl]-, 1,3-Benzenediol, 5-[2-(4-hydroxyphenyl)ethenyl]-, (Z)-, 5-[(1Z)-2-(4-Hydroxyphenyl)ethenyl]-1,3-benzenediol, (Z)-Resveratrol, cis-Resveratrol
IUPAC-namn 5-[(Z)-2-(4-hydroxyphenyl)ethenyl]benzene-1,3-diol
LEDER Oc1ccc(\C=C/c2cc(O)cc(O)c2)cc1
Specifikationer
Kemiskt namn eller material cis Resveratrol
Anteckningar om renhetsgrad HPLC
Kvantitet 5 mg
InChI formel InChI=1S/C14H12O3/c15-12-5-3-10(4-6-12)1-2-11-7-13(16)9-14(17)8-11/h1-9,15-17H/b2-1-
Formel vikt 228.0786
Procent renhet >95%