Läs mer
cis-trans Nepetalactone, TRC
Produktkod. 30010973
Change view
Kvantitet:
250 mg
2.5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30010973 2.5 g
30006210 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30010973 Leverantör Toronto Research Chemicals Leverantörsnummer TRCN3900852.5G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 21651-62-7
Molekylformel C10 H14 O2
Molekylvikt (g/mol) 166.22
Synonym Cyclopenta[c]pyran-1(4aH)-one, 5,6,7,7a-tetrahydro-4,7-dimethyl-, (4aS,7S,7aR)-, Cyclopenta[c]pyran-1(4aH)-one, 5,6,7,7a-tetrahydro-4,7-dimethyl-, [4aS-(4aα,7α,7aα)]-, Cyclopenta[c]pyran-1(4aH)-one, 5,6,7,7a-tetrahydro-4,7α-dimethyl-, (+)- (8CI), (4aS,7S,7aR)-5,6,7,7a-Tetrahydro-4,7-dimethylcyclopenta[c]pyran-1(4aH)-one, (+)-(4AS,7S,7aR)-nepetalactone, (+)-(4aS,7S,7aR)-Nepetalactone, (+)-cis,trans-Nepetalactone, (4aS,7S,7aR)-Nepetalactone, 4aα,7α,7aα-Nepetalactone, Nepetalactone, cis-trans-, cis-Trans nepetalactone, cis-trans-Nepetalactone
IUPAC-namn 4,7-dimethyl-5,6,7,7a-tetrahydro-4aH-cyclopenta[c]pyran-1-one
LEDER CC1CCC2C1C(=O)OC=C2C
Specifikationer
Kemiskt namn eller material cis-trans Nepetalactone
Anteckningar om renhetsgrad HPLC
Kvantitet 2.5 g
InChI formel InChI=1S/C10H14O2/c1-6-3-4-8-7(2)5-12-10(11)9(6)8/h5-6,8-9H,3-4H2,1-2H3
Formel vikt 166.0994
Procent renhet >95%