Läs mer
cis,trans-Sorbic Acid, TRC
Produktkod. 30009851
Change view
Kvantitet:
25 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30009851 100 mg
30006637 25 mg
30008877 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30009851 Leverantör Toronto Research Chemicals Leverantörsnummer TRCS676898100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 5309-57-9
Molekylformel C6H8O2
Molekylvikt (g/mol) 112.13
Synonym (2Z,4E)-2,4-Hexadienoic acid, 2-cis,4-trans-Hexadienoic acid
IUPAC-namn (2Z,4E)-hexa-2,4-dienoic acid
LEDER C\C=C\C=C/C(=O)O
Specifikationer
Kemiskt namn eller material cis,trans-Sorbic Acid
Kvantitet 100 mg
InChI formel InChI=1S/C6H8O2/c1-2-3-4-5-6(7)8/h2-5H,1H3,(H,7,8)/b3-2+,5-4-
Formel vikt 112.0524