Läs mer
Cisapride, TRC
Produktkod. 30358313
Change view
Kvantitet:
50 mg
Produktkod. Kvantitet
30358313 50 mg
1 options
Produktkod. 30358313 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC49640050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Cisapride
CAS 81098-60-4
Molekylformel C23 H29 Cl F N3 O4
Kvantitet 50 mg
Synonym Benzamide, 4-amino-5-chloro-N-[1-[(3R,4S)-3-(4-fluorophenoxy)propyl]-3-methoxy-4-piperidinyl]-2-methoxy-, rel-, Benzamide, 4-amino-5-chloro-N-[1-[3-(4-fluorophenoxy)propyl]-3-methoxy-4-piperidinyl]-2-methoxy-, cis-, rel-4-Amino-5-chloro-N-[1-[(3R,4S)-3-(4-fluorophenoxy)propyl]-3-methoxy-4-piperidinyl]-2-methoxybenzamide, (±)-Cisapride, Acenalin, Alimix, Cipril, Cisad, Cisapride, Coordinax, Prepulsid, Propulsid, Propulsin, R 51619, Risamal, T 1341, cis-2-Methoxy-4-amino-5-chloro-N-[1-[3-(4-fluorophenoxy)propyl]-3-methoxy-4-piperidinyl]benzamide
InChI formel InChI=1S/C23H29ClFN3O4/c1-30-21-13-19(26)18(24)12-17(21)23(29)27-20-8-10-28(14-22(20)31-2)9-3-11-32-16-6-4-15(25)5-7-16/h4-7,12-13,20,22H,3,8-11,14,26H2,1-2H3,(H,27,29)/t20-,22+/m0/s1
LEDER CO[C@@H]1CN(CCCOc2ccc(F)cc2)CC[C@@H]1NC(=O)c3cc(Cl)c(N)cc3OC
IUPAC-namn 4-amino-5-chloro-N-[(3R,4S)-1-[3-(4-fluorophenoxy)propyl]-3-methoxypiperidin-4-yl]-2-methoxybenzamide
Molekylvikt (g/mol) 465.95
Formel vikt 465.1831