Läs mer
Citalopram Hydrochloride, TRC
Produktkod. 30360742
Change view
Kvantitet:
50 mg
Produktkod. Kvantitet
30360742 50 mg
1 options
Produktkod. 30360742 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC50482050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Citalopram Hydrochloride
CAS 85118-27-0
Molekylformel C20 H21 F N2 O . Cl H
Kvantitet 50 mg
Synonym 5-Isobenzofurancarbonitrile, 1-[3-(dimethylamino)propyl]-1-(4-fluorophenyl)-1,3-dihydro-, hydrochloride (1:1), 5-Isobenzofurancarbonitrile, 1-[3-(dimethylamino)propyl]-1-(4-fluorophenyl)-1,3-dihydro-, monohydrochloride (9CI), (±)-Citalopram hydrochloride, 1-[3-(Dimethylamino)propyl]-1-(4-fluorophenyl)-1,3-dihydroisobenzofuran-5-carbonitrile monohydrochloride, Citalopram hydrochloride, (1RS)-1-[3-(Dimethylamino)propyl]-1-(4-fluorophenyl)-1,3-dihydroisobenzofuran-5-carbonitrile hydrochloride
InChI formel InChI=1S/C20H21FN2O.ClH/c1-23(2)11-3-10-20(17-5-7-18(21)8-6-17)19-9-4-15(13-22)12-16(19)14-24-20;/h4-9,12H,3,10-11,14H2,1-2H3;1H
LEDER Cl.CN(C)CCCC1(OCc2cc(ccc12)C#N)c3ccc(F)cc3
IUPAC-namn 1-[3-(dimethylamino)propyl]-1-(4-fluorophenyl)-3H-2-benzofuran-5-carbonitrile;hydrochloride
Molekylvikt (g/mol) 360.85
Formel vikt 360.1405