missing translation for 'onlineSavingsMsg'
Läs mer

CITCO, TRC

Produktkod. 30368227
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
1 mg
10 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30368227 50 mg
30367547 10 mg
30359911 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 30368227 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC43304850MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 338404-52-7
Molekylformel C19H12Cl3N3OS
Molekylvikt (g/mol) 436.74
Synonym 6-(4-Chlorophenyl)imidazo[2,1-b]thiazole-5-carboxaldehyde O-[(3,4-Dichlorophenyl)methyl]oxime
IUPAC-namn 1-[6-(4-chlorophenyl)imidazo[2,1-b][1,3]thiazol-5-yl]-N-[(3,4-dichlorophenyl)methoxy]methanimine
LEDER ClC1=CC=C(C2=C(/C=N/OCC3=CC=C(Cl)C(Cl)=C3)N4C(SC=C4)=N2)C=C1

Specifications

Kemiskt namn eller material CITCO
Kvantitet 50 mg
InChI formel InChI=1S/C19H12Cl3N3OS/c20-14-4-2-13(3-5-14)18-17(25-7-8-27-19(25)24-18)10-23-26-11-12-1-6-15(21)16(22)9-12/h1-10H,11H2
Formel vikt 434.9767
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.