Learn More
Citicoline sodium salt, 97%, BLD Pharm™
Product Code. 30968244
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
1 g
5 g
10 g
25 g
100 g
500 g
Unit Size:
100g
100mg
10g
25g
500g
5g
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30968244 1 g -
30955373 10 g 10g
30960719 100 mg 100mg
30960720 25 g 25g
30958770 5 g 5g
30960724 100 g 100g
30967010 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Product Code. 30968244 Supplier BLD Pharm Supplier No. BD1218061g
 Restricted Item
Beräknad leverans: 27-11-2026
Specifications
CAS 33818-15-4
Molecular Formula C14H25N4NaO11P2
MDL Number MFCD00037486
Quantity 1 g
SMILES O[C@H]1[C@@H](O[C@@H]([C@H]1O)COP(OP(OCC[N+](C)(C)C)([O-])=O)([O-])=O)N2C(N=C(C=C2)N)=O.[Na+]
Molecular Weight (g/mol) 510.31
Percent Purity 97%
Chemical Name or Material Citicoline sodium salt